Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxyisophthalic acid 98%

Supplier: AMBEED, INC
Warning

A403129-100G A403129-25G A403129-5G
77003-942EA 60 USD InStock
77003-942 77003-944 77003-946
4-Methoxyisophthalic acid 98%
4-Methoxyisophthalic acid
Precaución: for Research Use Only


Formula: C₉H₈O₅
MDL Number: MFCD06208422
CAS Number: 2206-43-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty