Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

XL019 ≥98%

Supplier: AMBEED, INC
Warning

A403201-10MG A403201-1MG A403201-25MG A403201-2MG A403201-5MG
76990-250EA 51 USD InStock
76990-250 76990-252 76990-254 76990-256 76990-258
XL019 ≥98%
XL019
Precaución: for Research Use Only


Formula: C₂₅H₂₈N₆O₂
MDL Number: MFCD24386874
CAS Number: 945755-56-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty