Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-2-Aminocyclopentanol 97%, Ambeed.Inc

Supplier: AMBEED, INC
Danger

A403690-100MG A403690-1G A403690-250MG A403690-5G A403690-100G A403690-25G
76990-260EA 10 USD InStock
76990-260 76990-262 76990-264 76990-266 77684-139 77684-140
trans-2-Aminocyclopentanol 97%, Ambeed.Inc
trans-2-Aminocyclopentanol
Precaución: For research use only.


Formula: C₅H₁₁NO
MDL Number: MFCD08669809
CAS Number: 59260-76-3
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty