Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-3-chlorobenzene-1,2-diamine 97%

Supplier: AMBEED, INC
Warning

A404956-10G A404956-1G A404956-5G
76990-276EA 89 USD InStock
76990-276 76990-278 76990-280
5-Bromo-3-chlorobenzene-1,2-diamine 97%
5-Bromo-3-chlorobenzene-1,2-diamine
Precaución: for Research Use Only


Formula: C₆H₆BrClN₂
MDL Number: MFCD12028675
CAS Number: 16429-44-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty