Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-(tert-Butylthio)phenyl)boronic acid 98%

Supplier: AMBEED, INC
Warning

A406046-100MG A406046-1G A406046-250MG A406046-5G
77016-328EA 13 USD InStock
77016-328 77016-330 77016-332 77016-334
(4-(tert-Butylthio)phenyl)boronic acid 98%
(4-(tert-Butylthio)phenyl)boronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₅BO₂S
MW: 210.10 g/mol
MDL Number: MFCD06801702
CAS Number: 820972-68-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty