Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dibromothiophene-3,4-dicarboxylic acid 95%

Supplier: AMBEED, INC
Warning

A406526-100MG A406526-1G A406526-250MG A406526-25G A406526-5G
77016-336EA 8 USD InStock
77016-336 77016-338 77016-340 77016-342 77016-344
2,5-Dibromothiophene-3,4-dicarboxylic acid 95%
2,5-Dibromothiophene-3,4-dicarboxylic acid
Precaución: for Research Use Only


Formula: C₆H₂Br₂O₄S
MW: 329.95 g/mol
MDL Number: MFCD08695787
CAS Number: 190723-12-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty