Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrafluorotetracyanoquinodimethane 98%

Supplier: AMBEED, INC
Danger

A407450-100MG A407450-1G A407450-250MG A407450-5G
77003-990EA 14 USD InStock
77003-990 77003-992 77594-234 77672-986
Tetrafluorotetracyanoquinodimethane 98%
Tetrafluorotetracyanoquinodimethane
Precaución: for Research Use Only


Formula: C₁₂F₄N₄
MDL Number: MFCD00042382
CAS Number: 29261-33-4
UN: 3439
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty