Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Hydroxyacetyl)benzonitrile 95%

Supplier: AMBEED, INC
Warning

A409104-100MG A409104-1G A409104-250MG
77016-354EA 20 USD InStock
77016-354 77016-356 77016-358
4-(2-Hydroxyacetyl)benzonitrile 95%
4-(2-Hydroxyacetyl)benzonitrile
Precaución: for Research Use Only


Formula: C₉H₇NO₂
MDL Number: MFCD00228206
CAS Number: 36776-32-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty