Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl (trans-4-aminocyclohexyl)carbamate hydrochloride 98%

Supplier: AMBEED, INC
Warning

A409138-1G A409138-250MG
76990-306EA 19 USD InStock
76990-306 76990-308
Benzyl (trans-4-aminocyclohexyl)carbamate hydrochloride 98%
Benzyl (trans-4-aminocyclohexyl)carbamate hydrochloride


Formula: C₁₄H₂₁ClN₂O₂
MDL Number: MFCD11007681
CAS Number: 1217664-37-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty