Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-N,N-Diethylpyrrolidin-3-amine dihydrochloride 95%

Supplier: AMBEED, INC
Warning

A413323-100MG A413323-1G A413323-250MG
76990-352EA 119 USD InStock
76990-352 76990-354 76990-356
(R)-N,N-Diethylpyrrolidin-3-amine dihydrochloride 95%
(R)-N,N-Diethylpyrrolidin-3-amine dihydrochloride
Precaución: for Research Use Only


Formula: C₈H₂₀Cl₂N₂
MDL Number: MFCD26395232
CAS Number: 1181083-20-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty