Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-D-2-cyanophenylalanine 95%

Supplier: AMBEED, INC
Warning

A417263-100MG A417263-1G A417263-250MG
77004-064EA 40 USD InStock
77004-064 77004-066 77004-068
Boc-D-2-cyanophenylalanine 95%
Boc-D-2-cyanophenylalanine
Precaución: for Research Use Only


Formula: C₁₅H₁₈N₂O₄
MDL Number: MFCD01862940
CAS Number: 261380-28-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty