Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloropyrimidine-2,4,5-triamine 97%

Supplier: AMBEED, INC
Warning

A419105-1G A419105-250MG
77004-092EA 246 USD InStock
77004-092 77004-094
6-Chloropyrimidine-2,4,5-triamine 97%
6-Chloropyrimidine-2,4,5-triamine
Precaución: for Research Use Only


Formula: C₄H₆ClN₅
MDL Number: MFCD11114448
CAS Number: 1194-78-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty