Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GSK2126458 ≥98%

Supplier: AMBEED, INC
Warning

A419303-10MG A419303-1MG A419303-2MG A419303-5MG
77033-836EA 80 USD InStock
77033-836 77033-838 77033-840 77033-842
GSK2126458 ≥98%
GSK2126458
Precaución: for Research Use Only


Formula: C₂₅H₁₇F₂N₅O₃S
MW: 505.5 g/mol
MDL Number: MFCD16038929
CAS Number: 1086062-66-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty