Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-6-fluoronicotinic acid 98%

Supplier: AMBEED, INC
Warning

A419466-100MG A419466-1G A419466-250MG A419466-5G
77033-844EA 46 USD InStock
77033-844 77033-846 77033-848 77033-850
5-Bromo-6-fluoronicotinic acid 98%
5-Bromo-6-fluoronicotinic acid
Precaución: for Research Use Only


Formula: C₆H₃BrFNO₂
MW: 219.996 g/mol
MDL Number: MFCD11036154
CAS Number: 29241-63-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty