Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Alogliptin benzoate 97%

Supplier: AMBEED, INC
Warning

A420160-100MG A420160-50MG A420160-25G A420160-5G
77033-858EA 8 USD InStock
77033-858 77033-860 77668-884 77668-886
Alogliptin benzoate 97%
Alogliptin benzoate
Precaución: for Research Use Only


Formula: C₁₈H₂₁N₅O₂·C₇H₆O₂
MW: 461.52 g/mol
MDL Number: MFCD09833195
CAS Number: 850649-62-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty