Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium ethynyltrifluoroborate 95%

Supplier: AMBEED, INC
Warning

A421718-100MG A421718-1G A421718-5G A421718-250MG
77033-874EA 17 USD InStock
77033-874 77033-876 77033-878 77668-888
Potassium ethynyltrifluoroborate 95%
Potassium ethynyltrifluoroborate
Precaución: for Research Use Only


Formula: C₂HBF₃K
MDL Number: MFCD10700167
CAS Number: 892869-21-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty