Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dimethoxythiophenol 97%

Supplier: AMBEED, INC
Danger

A422597-100MG A422597-250MG A422597-25G A422597-5G
77004-146EA 45 USD InStock
77004-146 77004-148 77004-150 77004-152
2,4-Dimethoxythiophenol 97%
2,4-Dimethoxythiophenol
Precaución: for Research Use Only


Formula: C₈H₁₀O₂S
MDL Number: MFCD04973436
CAS Number: 18906-37-1
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty