Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-tert-Butyl-4-nitrobenzenesulphonamide 98%

Supplier: AMBEED, INC
Warning

A423688-10G A423688-1G A423688-5G
77004-164EA 100 USD InStock
77004-164 77004-166 77004-168
N-tert-Butyl-4-nitrobenzenesulphonamide 98%
N-tert-Butyl-4-nitrobenzenesulphonamide
Precaución: for Research Use Only


Formula: C₁₀H₁₄N₂O₄S
MW: 258.29 g/mol
MDL Number: MFCD00443679
CAS Number: 49690-09-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty