Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinate 95%

Supplier: AMBEED, INC
Warning

A426071-100MG A426071-250MG A426071-1G
77033-888EA 30 USD InStock
77033-888 77033-890 77668-900
Methyl 2-amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinate 95%
Methyl 2-amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinate
Precaución: for Research Use Only


Formula: C₁₃H₁₉BN₂O₄
MW: 278.11 g/mol
MDL Number: MFCD12923412
CAS Number: 947249-44-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty