Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-4-methoxybenzenecarbohydrazide 95%

Supplier: AMBEED, INC
Warning

A426577-100MG A426577-1G A426577-250MG A426577-5G
77004-178EA 6 USD InStock
77004-178 77004-180 77004-182 77004-184
2-Hydroxy-4-methoxybenzenecarbohydrazide 95%
2-Hydroxy-4-methoxybenzenecarbohydrazide
Precaución: for Research Use Only


Formula: C₈H₁₀N₂O₃
MDL Number: MFCD06797386
CAS Number: 41697-08-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty