Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-(2-hydroxyethyl)thiophene-2-carboxylate 95%

Supplier: AMBEED, INC
Warning

A428602-10G A428602-1G A428602-250MG A428602-5G
77033-906EA 989 USD InStock
77033-906 77033-908 77033-910 77033-912
Methyl 5-(2-hydroxyethyl)thiophene-2-carboxylate 95%
Methyl 5-(2-hydroxyethyl)thiophene-2-carboxylate
Precaución: for Research Use Only


Formula: C₈H₁₀O₃S
MDL Number: MFCD12911672
CAS Number: 160744-13-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty