Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Quinoxaline-6-acetic acid 95%

Supplier: AMBEED, INC
Warning

A432730-100MG A432730-1G
77033-940EA 84 USD InStock
77033-940 77033-942
Quinoxaline-6-acetic acid 95%
Quinoxaline-6-acetic acid
Precaución: for Research Use Only


Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
MDL Number: MFCD09880609
CAS Number: 473932-16-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty