Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Pyridin-3-ylmethylpiperidin-4-ylamine 98+%

Supplier: AMBEED, INC
Warning

A438159-100MG A438159-1G A438159-250MG A438159-5G
77004-252EA 20 USD InStock
77004-252 77004-254 77004-256 77004-258
1-Pyridin-3-ylmethylpiperidin-4-ylamine 98+%
1-Pyridin-3-ylmethylpiperidin-4-ylamine
Precaución: for Research Use Only


Formula: C₁₁H₁₇N₃
MDL Number: MFCD08059920
CAS Number: 160357-88-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty