Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Carbobenzoxy-D-2-phenylglycine 98%

Supplier: AMBEED, INC
Warning

A441133-100G A441133-1G A441133-25G A441133-5G
77004-260EA 55 USD InStock
77004-260 77004-262 77004-264 77004-266
N-Carbobenzoxy-D-2-phenylglycine 98%
N-Carbobenzoxy-D-2-phenylglycine
Precaución: for Research Use Only


Formula: C₁₆H₁₅NO₄
MW: 285.29 g/mol
MDL Number: MFCD00021703
CAS Number: 17609-52-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty