Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-fluoro-2-propoxyphenylboronic acid 95%

Supplier: AMBEED, INC
Warning

A445371-1G A445371-250MG A445371-5G
77016-708EA 65 USD InStock
77016-708 77016-710 77016-712
3-Bromo-5-fluoro-2-propoxyphenylboronic acid 95%
3-Bromo-5-fluoro-2-propoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₁BBrFO₃
MW: 276.895 g/mol
MDL Number: MFCD07369707
CAS Number: 868272-84-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty