Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-Benzoylcarbazole 98%

Supplier: AMBEED, INC
Warning

A447417-100MG A447417-1G A447417-250MG A447417-5G
77034-068EA 9 USD InStock
77034-068 77034-070 77034-072 77034-074
9-Benzoylcarbazole 98%
9-Benzoylcarbazole
Precaución: for Research Use Only


Formula: C₁₉H₁₃NO
MDL Number: MFCD00225468
CAS Number: 19264-68-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty