Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-6-fluoro-1H-indole 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A449458-100MG A449458-250MG A449458-1G
77016-746EA 32 USD InStock
77016-746 77016-748 77684-231
4-Chloro-6-fluoro-1H-indole 95%, Ambeed.Inc
4-Chloro-6-fluoro-1H-indole
Precaución: For research use only.


Formula: C₈H₅ClFN
MDL Number: MFCD07781557
CAS Number: 885520-79-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty