Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-N-methylacetamide 95%

Supplier: AMBEED, INC
Warning

A450000-100G A450000-1G A450000-250MG A450000-25G A450000-5G
77034-090EA 264 USD InStock
77034-090 77034-092 77034-094 77034-096 77034-098
2-Bromo-N-methylacetamide 95%
2-Bromo-N-methylacetamide
Precaución: for Research Use Only


Formula: C₃H₆BrNO
MDL Number: MFCD06800288
CAS Number: 34680-81-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty