Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-4-morpholinoaniline 95%

Supplier: AMBEED, INC
Warning

A452101-1G A452101-250MG A452101-25G A452101-5G
77004-320EA 15 USD InStock
77004-320 77004-322 77673-124 77673-126
2-Methoxy-4-morpholinoaniline 95%
2-Methoxy-4-morpholinoaniline
Precaución: for Research Use Only


Formula: C₁₁H₁₆N₂O₂
MW: 208.26 g/mol
MDL Number: MFCD09972210
CAS Number: 209960-91-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty