Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 5-chloropentanoate 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A452693-5G A452693-25G
76990-758EA 375 USD InStock
76990-758 77684-238
Ethyl 5-chloropentanoate 97%, Ambeed.Inc
Ethyl 5-chloropentanoate
Precaución: For research use only.


Formula: C₇H₁₃ClO₂
MDL Number: MFCD00045312
CAS Number: 2323-81-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty