Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-2-nitrobenzeneboronic acid 95%

Supplier: AMBEED, INC
Warning

A465278-1G A465278-250MG A465278-5G
77004-444EA 103 USD InStock
77004-444 77004-446 77004-448
5-Fluoro-2-nitrobenzeneboronic acid 95%
5-Fluoro-2-nitrobenzeneboronic acid
Precaución: for Research Use Only


Formula: C₆H₅BFNO₄
MW: 184.92 g/mol
MDL Number: MFCD09037802
CAS Number: 1329171-65-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty