Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-methyl-1,3,4-thiadiazole 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A468319-100MG A468319-250MG A468319-1G
77016-908EA 25 USD InStock
77016-908 77016-910 77684-258
2-Chloro-5-methyl-1,3,4-thiadiazole 95%, Ambeed.Inc
2-Chloro-5-methyl-1,3,4-thiadiazole
Precaución: For research use only.


Formula: C₃H₃ClN₂S
MDL Number: MFCD17170011
CAS Number: 53645-94-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty