Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethylamino-p-tolylacetic acid 97%

Supplier: AMBEED, INC
Warning

A470532-100MG A470532-1G A470532-250MG
77004-466EA 67 USD InStock
77004-466 77004-468 77004-470
Dimethylamino-p-tolylacetic acid 97%
Dimethylamino-p-tolylacetic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₅NO₂
MDL Number: MFCD08059910
CAS Number: 230646-18-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty