Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl octanoate 98%

Supplier: AMBEED, INC
Warning

A472307-100G A472307-25G A472307-500G
77004-484EA 6 USD InStock
77004-484 77004-486 77004-488
Ethyl octanoate 98%
Ethyl octanoate
Precaución: for Research Use Only


Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 206
Melting Pt: -47
Flash Pt: 81
MDL Number: MFCD00009552
CAS Number: 106-32-1
Merck Index: 13,03813
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty