Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3-chloro-6-methylpyridine 95%

Supplier: AMBEED, INC
Warning

A472970-100MG A472970-1G A472970-250MG
76990-924EA 22 USD InStock
76990-924 76990-926 76990-928
2-Bromo-3-chloro-6-methylpyridine 95%
2-Bromo-3-chloro-6-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₅BrClN
MDL Number: MFCD13185356
CAS Number: 1211582-97-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty