Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dimethyl-1H-pyrazole-4-sulfonyl chloride 98%

Supplier: AMBEED, INC
Danger

A473262-25G A473262-5G A473262-250MG
77034-300EA 690 USD InStock
77034-300 77034-302 77673-224
1,3-Dimethyl-1H-pyrazole-4-sulfonyl chloride 98%
1,3-Dimethyl-1H-pyrazole-4-sulfonyl chloride
Precaución: for Research Use Only


Formula: C₅H₇ClN₂O₂S
MDL Number: MFCD03944999
CAS Number: 89501-93-9
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty