Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl-3,3'-iminodipropionate 98%

Supplier: AMBEED, INC
Warning

A474093-250MG A474093-5G
77016-966EA 31 USD InStock
77016-966 77016-968
Diethyl-3,3'-iminodipropionate 98%
Diethyl-3,3'-iminodipropionate
Precaución: for Research Use Only


Formula: C₁₀H₁₉NO₄
MW: 217.27 g/mol
MDL Number: MFCD14705114
CAS Number: 3518-88-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty