Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2-chloronicotinaldehyde 95%

Supplier: AMBEED, INC
Warning

A474333-100MG A474333-1G A474333-250MG
77034-304EA 22 USD InStock
77034-304 77034-306 77034-308
4-Amino-2-chloronicotinaldehyde 95%
4-Amino-2-chloronicotinaldehyde
Precaución: For research use only.


Formula: C₆H₅ClN₂O
MDL Number: MFCD09834659
CAS Number: 338452-92-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty