Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-m-anisaldehyde 98%

Supplier: AMBEED, INC
Warning

A476397-100G A476397-10G A476397-1G A476397-25G A476397-5G
77004-516EA 263 USD InStock
77004-516 77004-518 77004-520 77004-522 77004-524
4-Fluoro-m-anisaldehyde 98%
4-Fluoro-m-anisaldehyde
Precaución: for Research Use Only


Formula: C₈H₇FO₂
MDL Number: MFCD00143320
CAS Number: 128495-46-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty