Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-iodoisonicotinate 96%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A478285-100MG A478285-250MG A478285-1G
77016-996EA 17 USD InStock
77016-996 77016-998 77684-272
Methyl-2-iodoisonicotinate 96%, Ambeed.Inc
Methyl 2-iodoisonicotinate
Precaución: For research use only.


Formula: C₇H₆INO₂
MDL Number: MFCD08235123
CAS Number: 134579-47-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty