Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromoimidazo[1,2-a]pyridine-2-carbaldehyde 96%

Supplier: AMBEED, INC
Warning

A482190-100MG A482190-1G A482190-250MG
77017-032EA 37 USD InStock
77017-032 77017-034 77017-036
6-Bromoimidazo[1,2-a]pyridine-2-carbaldehyde 96%
6-Bromoimidazo[1,2-a]pyridine-2-carbaldehyde
Precaución: for Research Use Only


Formula: C₈H₅BrN₂O
MW: 225.04 g/mol
MDL Number: MFCD06739238
CAS Number: 885276-09-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty