Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-2-(4-octylphenethyl)propane-1,3-diol 98%

Supplier: AMBEED, INC
Warning

A484670-10MG A484670-1MG A484670-250MG
77034-372EA 24 USD InStock
77034-372 77034-374 77034-376
2-Amino-2-(4-octylphenethyl)propane-1,3-diol 98%
2-Amino-2-(4-octylphenethyl)propane-1,3-diol
Precaución: for Research Use Only


Formula: C₁₉H₃₃NO₂
MDL Number: MFCD14705150
CAS Number: 162359-55-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty