Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glycine benzyl ester-p-toluenesulfonate 98%

Supplier: AMBEED, INC
Warning

A487822-100G A487822-10G A487822-5G A487822-500G
77004-608EA 60 USD InStock
77004-608 77004-610 77004-612 77673-272
Glycine benzyl ester-p-toluenesulfonate 98%
Glycine benzyl ester-p-toluenesulfonate
Precaución: for Research Use Only


Formula: C₉H₁₁NO₂
MDL Number: MFCD00035425
CAS Number: 1738-76-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty