Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-2-Phenylethenesulfonyl chloride 97%

Supplier: AMBEED, INC
Danger

A491072-10G A491072-250MG A491072-25G
77017-138EA 122 USD InStock
77017-138 77017-140 77017-142
(E)-2-Phenylethenesulfonyl chloride 97%
(E)-2-Phenylethenesulfonyl chloride
Precaución: For research use only.


Formula: C₈H₇ClO₂S
MDL Number: MFCD00007456
CAS Number: 52147-97-4
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty