Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trifluoromethyl)-o-anisidine 98%

Supplier: AMBEED, INC
Danger

A491109-100MG A491109-1G A491109-250MG A491109-5G
76991-114EA 10 USD InStock
76991-114 76991-116 76991-118 76991-120
3-(Trifluoromethyl)-o-anisidine 98%
3-(Trifluoromethyl)-o-anisidine
Precaución: for Research Use Only


Formula: C₈H₈F₃NO
MDL Number: MFCD08532489
CAS Number: 634187-08-9
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty