Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3'-(5'-(4-(Pyridin-3-yl)phenyl)-[1,1':3',1''-terphenyl]-4,4''-diyl)dipyridine 95+%

Supplier: AMBEED, INC
Warning

A492073-100MG A492073-1G A492073-250MG A492073-5G
77017-158EA 50 USD InStock
77017-158 77017-160 77017-162 77017-164
3,3'-(5'-(4-(Pyridin-3-yl)phenyl)-[1,1':3',1''-terphenyl]-4,4''-diyl)dipyridine 95+%
3,3'-(5'-(4-(Pyridin-3-yl)phenyl)-[1,1':3',1''-terphenyl]-4,4''-diyl)dipyridine
Precaución: for Research Use Only


Formula: C₃₉H₂₇N₃
MDL Number: MFCD27923078
CAS Number: 921205-02-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty