Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Difluoro-L-phenylalanine 95%

Supplier: AMBEED, INC
Warning

A493943-100MG A493943-1G A493943-250MG
77004-638EA 20 USD InStock
77004-638 77004-640 77004-642
2,5-Difluoro-L-phenylalanine 95%
2,5-Difluoro-L-phenylalanine
Precaución: for Research Use Only


Formula: C₉H₉F₂NO₂
MW: 201.17 g/mol
MDL Number: MFCD07372000
CAS Number: 31105-92-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty