Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Propoxypicolinimidamide hydrochloride 98%

Supplier: AMBEED, INC
Warning

A495711-1G A495711-250MG
76991-192EA 136 USD InStock
76991-192 76991-194
3-Propoxypicolinimidamide hydrochloride 98%
3-Propoxypicolinimidamide hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₄ClN₃O
MDL Number: MFCD12755577
CAS Number: 1179362-43-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty