Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-1-methyl-2,4-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole 95%

Supplier: AMBEED, INC
Warning

A497286-1G A497286-250MG A497286-5G
77017-200EA 209 USD InStock
77017-200 77017-202 77017-204
6-Chloro-1-methyl-2,4-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole 95%
6-Chloro-1-methyl-2,4-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
Precaución: for Research Use Only


Formula: C₂₁H₃₀B₂ClNO₄
MDL Number: MFCD17015835
CAS Number: 1256360-39-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty