Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Nitro-1H-indazol-6-ol 97%

Supplier: AMBEED, INC
Warning

A498154-100MG A498154-1G A498154-250MG
77017-206EA 10 USD InStock
77017-206 77017-208 77017-210
5-Nitro-1H-indazol-6-ol 97%
5-Nitro-1H-indazol-6-ol
Precaución: for Research Use Only


Formula: C₇H₅N₃O₃
MDL Number: MFCD11007883
CAS Number: 1082041-56-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty